Learn More
Gilson™ Corrosion Protection kit
Ideal protection for PIPETMAN P1000, PIPETMAN NEO P1000N and PIPETMAN F1000 while pipetting corrosive liquids, such as acids
Brand: Gilson™ F144570
31.99 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Optimize the repeatability of your results and maintain the smoothness of your piston stroke
- Avoid contamination and leaks caused by intensive pipetting of corrosive solutions
- Save time and money by prolonging the life of your PIPETMAN