Learn More
Invitrogen™ Cortisol Competitive Human ELISA Kit
Competitive ELISA
£546.00 - £3590.00
Specifications
Assay Sensitivity | 17.3 pg/mL |
---|---|
Description | Cortisol Competitive ELISA Kit |
Conjugate | HRP |
Detection Method | Fluorescence |
Product Type | ELISA Kit |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
15642299
|
Invitrogen™
EIAHCOR |
96 assays |
£546.00
96 tests |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
17815217
|
Invitrogen™
EIAHCORX10 |
10 x 96 Assays |
£3590.00
Pack of 10 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
Description
The Human Cortisol ELISA quantitates Cortisol in human serum,plasma,urine,saliva,dried fecal extracts or or cell culture medium.
Principle of the method
The Cortisol Competitive ELISA research-use-only kit is designed to quantitatively measure cortisol present in dried fecal extracts,saliva,urine,serum,plasma,and tissue culture media samples. Total cortisol is measured in extracted samples and in serum and plasma,and free cortisol in saliva and urine. A cortisol standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture mouse antibodies. A cortisol-peroxidase conjugate is added to the wells. The binding reaction is initiated by the addition of a monoclonal antibody to cortisol. After a 1-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound cortisol-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
Specifications
17.3 pg/mL | |
HRP | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
8.8% | |
RUO | |
-20°C | |
1 hr. 20 min. |
Cortisol Competitive ELISA Kit | |
Fluorescence | |
Fecal Extract, Plasma, Saliva, Serum, Supernatant, Urine | |
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti | |
8.1% | |
HRP | |
Inflammation | |
Human | |
1 hr. 30 min. |