missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 8-Well Polypropylene PCR Tube Strips

Product Code. 10348421 Shop All Corning Products
Click to view available options
Color:
Assorted
Clear
Unit Size:
Pack of 300
This item is not returnable. View return policy

Product Code. 10348421

Brand: Corning™ 6542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured from thin-walled polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Corning™ 8-Well PCR Tube Strips are color coded for easy sample identification.

Tube strips consist of eight 0.2 mL thin wall polypropylene tubes connected together. Dual connectors between adjacent tubes eliminate inadvertent breakage during sample handling. Tube strips are designed for precise fit in thermal cyclers to optimize heat transfer. Thermowell GOLD cap strips are sold separately from Thermowell GOLD tube strips. Original Thermowell tube strips and cap strips are packaged together. Tube strips are RNase-/DNase-free, and are autoclavable at 121°C.



Cat. No. 6547 has 60 of each color per bag; 1 bag of each color per case.

Cat. No. 3742 is designed for real-time PCR. Suitable for use with Thermowell GOLD 0.2 mL 1 x 8 PCR tube strips and 96-well microplates.

Order Info

Cap strips are packaged together with tube strips.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase-free
Color Clear
Volume (Metric) 0.2 mL
Quantity 60/Pk.
Closure Material Polypropylene
For Use With (Equipment) Heat blocks to optimize heat transfer
Shape Conical
Sterility Non-sterile
Material Polypropylene
Autoclavable Autoclavable
Format Strip
Product Type Thin Wall 8-Strip PCR Tube
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.