missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Assay Blocks
Shop All Corning Products

Click to view available options
Sterility:
Non-sterile
Sterile
Volume (Metric) Well:
500 μL
1000 μL
2000 μL
Well Shape:
Conical Bottom (V-well)
Round
Round, Conical Bottom (V-well)
No. of Wells:
96
Unit Size:
Pack of 100
Pack of 25
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Certifications
Certified DNase- and RNase-free
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Packaging | 5 inner bags of 5 each |
| Binding Type | Not Treated |
| Color | Clear |
| Product Type | 96-well Assay Blocks |
| Material | Polypropylene |
| Volume (Metric) Well | 1000 μL |
| No. of Wells | 96 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction