Learn More
Corning™ 96-Well Assay Blocks
Manufactured from clarified polypropylene for optical clarity. Corning™ 96-Well Assay Blocks are ideal for compound storage, assay, and cell-growth applications.
Marca: Corning™ 3959
Code : 66
Dettagli aggiuntivi : Weight : 11.14200kg
Descrizione
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Specifica
Standard | |
8 x 12 | |
Clear | |
1000 μL | |
Non-sterile | |
Round | |
100/Cs. |
Without Lid | |
Not Treated | |
Polypropylene | |
96 | |
Non-treated | |
For compound storage, assay and cell-growth application | |
5/Pk. |