Learn More
Corning™ 96-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Brand:  Corning™ 3915
404.82 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
 View more versions of this product
Product Code. 10022561
  • £501.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Black
    96
    Non-sterile
    Flat Bottom (F-well)
    Polystyrene
    75 to 200 μL
    Non-treated
    Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.