Learn More
Corning™ Clear Polystyrene 96-Well Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Brand: Corning™ 9017
Code : 66
Additional Details : Weight : 5.66820kg
Description
- Medical-grade polystyrene
- Well volume: 320μL to 360μL
- Working volume: 75μL to 200μL
- Choice of flat, round, flat Easy Wash™ or V-shaped bottom
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
- Plate dimensions meet proposed industry standards
Specifications
Medium Binding | |
Without Lid | |
Clear | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. |
Standard | |
8 x 12 | |
Polystyrene | |
75 to 200 μL | |
Non-treated | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. |