missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Clear PVC Assay Microplates
Manufactured from clear flexible Polyvinyl Chloride (PVC) for economy and visibility. Corning™ 96-Well Clear PVC Assay Microplates present an excellent solution for laboratory miniaturization. Due to their flexible nature, these microplates are not compatible with automation.
£208.13 - £222.33
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 8 x 12 |
Color | Clear |
Material | PVC |
Product Code | Brand | Well Shape | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Shape | Price | Quantity & Availability | |||||
10574201
|
Corning™
2797 |
Round |
£210.33
Pack of 100 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||||
10691432
|
Corning™
2595 |
Flat Bottom (F-well) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10639541
|
Corning™
2897 |
Conical Bottom (V-well) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Order Info
Lids are not available. Supplied in packs of 25.
Specifications
Standard | |
8 x 12 | |
PVC | |
50 to 150 μL | |
Non-treated | |
100/Cs. | |
100/Cs. |
Without Lid | |
Clear | |
96 | |
Non-sterile | |
Solution-based assays, serial dilutions and general storage applications | |
25/Pk. |