Learn More
Corning™ 96-Well Half-Area Plates

Saves on valuable reagents by reducing the amount of reagent required per well, while retaining the ability to be read in standard microplate readers. Corning™ 96-Well Half Area Microplates are an economical solution for all of your assay diagnostic needs and are available in different colors and treatments.
Brand: Corning™ 3686
1159.00 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description

Specifications
Alphanumeric | |
Without Lid | |
4 inner packs of 25 | |
Plate readers | |
190 μL | |
25 to 125 μL | |
Non-binding | |
Minimization of protein and nucleic acid binding | |
25/Pk. | |
Black |
Half-Area | |
8 x 12 | |
Black | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
10Hours
22Minutes
3Seconds
0
Your input is important to us. Please complete this form to provide feedback related to the content on this product.