missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate
Shop All Corning Products

Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Specifications
| Barcode | Alphanumeric |
| Format | Half-Area |
| Lid | Without Lid |
| Array | 8 x 12 |
| Binding Type | Not Treated |
| Color | Clear |
| For Use With (Equipment) | Plate readers |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 25 to 125 μL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction