Learn More
Corning™ Aspirator
Ideal for removal of supernatants - an alternative to glass Pasteur pipets
Brand: Corning™ 4930
Includes: Handpiece, grommet for attachment of accessories, and a single-channel adapter for use with disposable pipetter tips.
Description
The Corning™ Aspirator is a device using vacuum to safely and aseptically dispose of supernatant from a variety of laboratory vessels. The aspirator includes a hand piece to which disposable pipet tips are attached during use, a grommet for accessory attachment and single-channel adapter for disposable pipet tips.
An 8-channel adapter is available for working in 96 well plates, see No. 4931. Refer to the accessories tab below.
Specifications
Autoclavable | |
Aspirator | |
Disposable pipette tip, Polyethylene and Polystyrene aspiration pipette, or Pasteur pipette |
Costar | |
Safe removal/disposal of supernatants from a wide range of laboratory vessels | |
Handpiece, grommet for accessory attachment and single channel adapter for disposable pipette tip |