missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 6-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
GREENER_CHOICEBIOPHARMA

Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.

Brand:  Corning™ 3516

131.25 GBP valid until 2025-03-31
Use promo code "24073" to get your promotional price.


Product Code. 10578911

  • £156.00 / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 x 3
Yes
Flat
Clear
Polystyrene
Sterile
Multi-well Cell Culture Plate
6
50/Cs.
Individually Wrapped
3.5 cm
Clear
Polystyrene
2.9 mL (Well)
9.5 cm2
Rectangular
Cell Culture
2 to 3 mL
Lid
1/Pk.
50/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number34124014 Certificate TypeCertificate of Quality Date29/03/2025 Catalog Number3516
Lot Number33924045 Certificate TypeCertificate of Quality Date29/03/2025 Catalog Number3516
Lot Number33924007 Certificate TypeCertificate of Quality Date25/03/2025 Catalog Number3516
Lot Number33524002 Certificate TypeCertificate of Quality Date12/03/2025 Catalog Number3516
Lot Number33224004 Certificate TypeCertificate of Quality Date12/03/2025 Catalog Number3516
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Costar™ 6-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile > 6-well; TC-treated; 1/Pk; 50/Cs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.