missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, V-Shaped-Bottom Microplate
GREENER_CHOICE

Color:Clear; Sterility:Sterile; Lid:With Lid

Brand:  Corning™ 3894

163.89 GBP valid until 2025-03-31
Use promo code "24073" to get your promotional price.


Product Code. 10462012

  • £213.00 / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
0.38 cm2
TC-Treated
Polystyrene
75 to 200 μL
Cell Culture
Conical Bottom (V-well)
50/Cs.
50/Cs.
With Lid
8 x 12
Clear
96
Sterile
0.38 mL
Attachment and growth of anchorage-dependent cells
1/Pk.
1.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number31324046 Certificate TypeCertificate of Quality Date25/03/2025 Catalog Number3894
Lot Number33524010 Certificate TypeCertificate of Quality Date18/03/2025 Catalog Number3894
Lot Number28024004 Certificate TypeCertificate of Quality Date05/03/2025 Catalog Number3894
Lot Number27524032 Certificate TypeCertificate of Quality Date01/01/2025 Catalog Number3894
Lot Number17924034 Certificate TypeCertificate of Quality Date05/12/2024 Catalog Number3894
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

    New Offer
    Save Now
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    44

    Hours

    23

    Minutes

    49

    Seconds

    54
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ 96-Well, Cell Culture-Treated, V-Shaped-Bottom Microplate > Standard, with Lid; 1/Pk; Yes; TC-treated; V-bottom

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.