missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ 24-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile



Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
Brand: Corning™ 3526
Description
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic

Specifications
4 x 6 | |
Yes | |
Flat | |
Clear | |
Polystyrene | |
Sterile | |
Multi-well Cell Culture Plate | |
24 | |
50/Cs. | |
Individually Wrapped | |
1.6 cm |
Clear | |
Polystyrene | |
0.57 mL (Well) | |
1.9 cm2 | |
Rectangular | |
Cell Culture | |
0.5 mL | |
Lid | |
1/Pk. | |
50/Cs. |
COR001 Lifescience Guid
File size: 7 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
New Offer Save Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ Costar™ 24-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile > 24-well; TC-treated; 1/Pk; 50/Cs
Spot an opportunity for improvement?Share a Content Correction