missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ 6-well Clear TC-treated Multiple Well Plates, Bulk Packed, Sterile
GREENER_CHOICEBIOPHARMA

Product Code. 10146810 Shop All Corning Products
Click to view available options
Diameter (Metric):
3.5 cm
Unit Size:
Pack of 100

Product Code. 10146810

Brand: Corning™ 3506

Only null left
Add to basket
Edge
Add to basket

Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY

Specifications

Array 2 x 3
Closure Color Clear
Closure Included Yes
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 2.9 mL (Well)
Color Clear
Culture Area 9.5 cm2
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Cell Culture
Type Multi-well Cell Culture Plate
Volume (Metric) Working 2 to 3 mL
Well Count 6
Includes Lid
No. per Case 100/Cs.
No. per Pack 5/Pk.
Packaging Bulk
Quantity 100/Cs.
Diameter (Metric) 3.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.