missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Cell Culture Triangular Flasks
GREENER_CHOICE

Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.

£250.53 - £563.69

Specifications

Closure Material HDPE
Material Polystyrene
For Use With (Application) For optimal cell attachment and growth
Shape Triangular
Surface Coating Tissue Culture Treated
View More Specs

Products 4
Product Code Brand dimCultureArea Closure Color Neck Style VolumeMetricWorking Price Quantity & Availability  
Product Code Brand dimCultureArea Closure Color Neck Style VolumeMetricWorking Price Quantity & Availability  
10762122
GREENER_CHOICE
View Documents
Corning™
3276
Orange Straight This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10005240
GREENER_CHOICE
View Documents
Corning™
3056
Orange Angled This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10539442
GREENER_CHOICE
View Documents
Corning™
3055
Black Angled This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10466302
GREENER_CHOICE
View Documents
Corning™
3275
Black Straight This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

 

  • Medical-grade polystyrene is sterile and nonpyrogenic
  • Tissue culture-treated to promote optimal cell attachment and growth
  • Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
  • Graduated
  • Frosted labeling surfaces
  • Extra-wide neck openings for easier pipetting
  • Printed lot number and catalog number on each flask
  • Vented filter caps
  • Maximize gas exchange while minimizing the possibility of contamination
  • Hydrophobic 0.2 μm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened

 

Specifications

Specifications

HDPE
For optimal cell attachment and growth
Tissue Culture Treated
Sterile
Polystyrene
Triangular
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.