missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ Clear Multiple Well Plates
Untreated surface is hydrophobic in nature and binds bio-olecules through passive interactions. Corning™ Costar™ Clear Multiple Well Plates provide an ideal solution for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface.
£259.61 - £293.30
Specifications
Format | Standard |
---|---|
Lid | With Lid |
Binding Type | Medium Binding |
Color | Clear |
Material | Polystyrene |
Product Code | Brand | Culture Area | Format | Volume (Metric) Well | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Culture Area | Format | Volume (Metric) Well | Price | Quantity & Availability | |||
11399494
|
Corning™
3737 |
3.8 cm2 | Standard | 6.9 mL |
£293.30
Pack of 100 |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||
11309504
|
Corning™
3738 |
1.9 cm2 | Standard | 3.4 mL |
£266.84
Pack of 100 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
11389494
|
Corning™
3736 |
9.5 cm2 | Standard | 16.8 mL |
£259.61
Pack of 100 |
In Stock
Log in to see stock. |
|||
Description
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Specifications
Standard | |
Medium Binding | |
Polystyrene | |
Non-treated | |
100/Cs. |
With Lid | |
Clear | |
Sterile | |
Flat Bottom (F-well) | |
5/Pk. |