Learn More
Corning™ Clear Polystyrene 96-Well Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
Brand: Corning™ 3370
Code : 66
Additional Details : Weight : 6.78600kg
Descripción
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Especificaciones
Standard | |
8 x 12 | |
Not Treated | |
EIA/RIA | |
96 | |
Sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |
With Lid | |
5 inner bags of 20 | |
Clear | |
Polystyrene | |
75 to 200 μL | |
Non-treated | |
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions. | |
20/Pk. |