missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Cluster Tube System

Product Code. 10285471 Shop All Corning Products
Click to view available options
Product Type:
8-Strip Microplate Cluster Tube
Microplate Cluster Tube
Format:
Strip
Tube
Quantity:
12 Strips/Rack, 120 Strips/Cs.
12/Rack
96/Rack
Bulk
Bulk, 120/Bag
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 120
Pack of 960
This item is not returnable. View return policy

Product Code. 10285471

Brand: Corning™ 4410

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.

  • Composed of 96 polypropylene tubes in a standard microplate format.
  • Features 1.2 mL tubes that are available individually or in strips of eight tubes.
  • Polyethylene tube caps are available in 8-cap strips.
TRUSTED_SUSTAINABILITY

Specifications

Format Tube
Quantity 96/Rack
Autoclavable Autoclavable
Material Polypropylene
Product Type Microplate Cluster Tube
Sterility Non-sterile
No. per Pack 96/Pk.
No. per Case 960/Cs.
Color Translucent
Volume (Metric) 1 mL
Includes Rack
For Use With (Equipment) Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges
Volume (English) 0.033 oz.
Well Count 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.