Learn More
Corning™ 96-Well Cluster Tube System
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Brand: Corning™ 4411
199.27 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Includes: Rack of 96 tubes. (Caps are sold separately.)
Description
- Composed of 96 polypropylene tubes in a standard microplate format.
- Features 1.2 mL tubes that are available individually or in strips of eight tubes.
- Polyethylene tube caps are available in 8-cap strips
- Caps are sold separately; see Corning Polyethylene Caps for Cluster Tube System (Mfr. No. 4418)

Specifications
Tube | |
Autoclavable | |
Microplate Cluster Tube | |
96/Pk. | |
Translucent | |
Rack of 96 tubes. (Caps are sold separately.) | |
0.033 oz. |
96/Rack | |
Polypropylene | |
Sterile | |
960/Cs. | |
1 mL | |
Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges | |
96 |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.