missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 96-Well Cluster Tube System
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Brand: Corning™ 4412
This item is not returnable.
View return policy
Description
- Composed of 96 polypropylene tubes in a standard microplate format.
- Features 1.2 mL tubes that are available individually or in strips of eight tubes.
- Polyethylene tube caps are available in 8-cap strips.

Specifications
Strip | |
Autoclavable | |
8-Strip Microplate Cluster Tube | |
12 strips/rack | |
Translucent | |
Rack | |
0.033 oz. |
12 Strips/Rack, 120 Strips/Cs. | |
Polypropylene | |
Non-sterile | |
120 Strips/Cs. | |
1 mL | |
Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges | |
96 |
COR001 Lifescience Guid
File size: 7 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 96-Well Cluster Tube System > Rack; No; 8-Tube Strip
Spot an opportunity for improvement?Share a Content Correction