missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ HTS Transwell™ -96 Well Permeable Support System

Shop All Corning Products


Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
No. per Pack:
1/Pk.
4/Pk.
5/Pk.
Unit Size:
Each
Pack of 2
Pack of 5
Pack of 8
Description
- The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
- Choice of either polyester (PET) membrane (0.4 μm, 1.0 μm pore sizes) or polycarbonate (PC) membrane (0.4 μm, 3.0 μm, and 5.0 μm pore sizes)
- 0.143 cm2 membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
- Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access.
- Optimized for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
- The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabilizer to reduce the risk of spills during handling
- The receiver plate isolates each well to enable 96 individual assays.
- Sterile
- The HTS Transwell-96 systems (0.4 μm PC, 0.4 μm PET, and 1.0 μm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid.
- The HTS Transwell-96 plates (3.0 and 5.0 μm PC, 8.0 μm PET) are packaged with the 96-well insert plate in the 96-well receiver plate with lid. Reservoir plates may be purchased separately.
Spécification
Spécification
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.143 cm2 |
| Membrane | TC-treated polycarbonate |
| Array | 8 x 12 |
| Pore Size | 0.4 μm |
| Color | Clear |
| Material | Polycarbonate |
| No. of Wells | 96 |
| Volume (Metric) Working | 0.23 mL |
| Afficher plus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction