missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ HTS Transwell™ -96 Well Permeable Support System
Boasts membrane area for cell growth or migration of 0.143cm2. Corning™ HTS Transwell™ 96-Well Permeable Support System is a microplate support system for an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
£166.90 - £1608.07
Specifications
Format | Standard |
---|---|
Culture Area | 0.143 cm2 |
Array | 8 x 12 |
Color | Clear |
No. of Wells | 96 |
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. per Pack | Price | Quantity & Availability | ||||
10175562
|
Corning™
3391 |
0.4 μm | 5/Pk. |
£1608.07
Pack of 5 |
In Stock
Log in to see stock. |
||||
10625053
|
Corning™
3381 |
0.4 μm | 1/Pk. |
£338.59
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||||
10399242
|
Corning™
3392 |
1 μm | 5/Pk. |
£1608.07
Pack of 5 |
In Stock
Log in to see stock. |
||||
10043832
|
Corning™
3380 |
1 μm | 1/Pk. |
£338.59
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
10271593
|
Corning™
3385 |
3 μm | 1/Pk. |
£166.90
Pack of 2 |
In Stock
Log in to see stock. |
||||
10077792
|
Corning™
3386 |
3 μm | 4/Pk. |
£667.37
Pack of 8 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||||
10797023
|
Corning™
3387 |
5 μm | 4/Pk. |
£667.37
Pack of 8 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
10555673
|
Corning™
3388 |
5 μm | 1/Pk. |
£166.90
Pack of 2 |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
Description
- The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
- Choice of either polyester (PET) membrane (0.4 μm, 1.0 μm pore sizes) or polycarbonate (PC) membrane (0.4 μm, 3.0 μm, and 5.0 μm pore sizes)
- 0.143 cm2 membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
- Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access.
- Optimized for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
- The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabilizer to reduce the risk of spills during handling
- The receiver plate isolates each well to enable 96 individual assays.
- Sterile
- The HTS Transwell-96 systems (0.4 μm PC, 0.4 μm PET, and 1.0 μm PET) are packaged with the 96-well insert plate in a reservoir plate and includes the 96-well receiver plate with lid.
- The HTS Transwell-96 plates (3.0 and 5.0 μm PC, 8.0 μm PET) are packaged with the 96-well insert plate in the 96-well receiver plate with lid. Reservoir plates may be purchased separately.
Specifications
Standard | |
8 x 12 | |
96 | |
Sterile | |
4.26 mm |
0.143 cm2 | |
Clear | |
0.23 mL | |
Flat Bottom (F-well) |