Learn More
Corning™ Costar™ Netwell™ Inserts
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers.
Brand:  Corning™ 3478
Code : 67
Additional Details : Weight : 0.44910kg
Product Code. 10187381
  • £424.65 / Pack of 48
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
Netwell Inserts have polyester (PET) mesh bottoms attached to a polystyrene ring or housing. They are used as tissue carriers, supports and strainers for culture of small organs, tissue slices, or explants at the air-media interface. They can be used to coarse filter tissue homogenates, cell suspensions, or microcarriers. Accessories allow them to be used as a handy carrier for immunocytochemical staining of tissue culture slices.
Specifications
Specifications
3 x 4
Clear
Round
Cell Culture Insert
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
12/Pk.
440 μm
Yes
Polystyrene
Sterile
12
48/Cs.
4 inner packs of 12
48/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products