Learn More
Corning™ Costar™ Netwell™ Inserts
GREENER_CHOICE
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers.
Brand:  Corning™ 3477
Product Code. 10426651
  • £457.00 / Pack of 48
Estimated Shipment: 28-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Netwell Inserts have polyester (PET) mesh bottoms attached to a polystyrene ring or housing. They are used as tissue carriers, supports and strainers for culture of small organs, tissue slices, or explants at the air-media interface. They can be used to coarse filter tissue homogenates, cell suspensions, or microcarriers. Accessories allow them to be used as a handy carrier for immunocytochemical staining of tissue culture slices.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Used as tissue carriers, supports and strainers for culture of small organs, tissue slices or explants at the air media interface, provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
Polystyrene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Costar™ Netwell™ Inserts > 15 mm; 74 μm