Learn More
Corning™ Costar™ Sterile Disposable Reagent Reservoirs
Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.
Brand: Corning™ 4873
275.39 GBP valid until 2024-12-31
Use promo code "22009" to get your promotional price.
Description
Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.
Costar™ reagent reservoirs are ideal for repetitively filling multichannel pipettors.
- Manufactured from modified polystyrene (white)
- Individually wrapped, sterile
- Disposable
Specifications
100 mL | |
Polystyrene | |
35 mm | |
100/Cs. | |
Individually Wrapped | |
Single-use |
145 x 85 x 35 mm | |
Sterile | |
145 mm | |
1/Pk. | |
88.5 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.