missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Sterile Disposable Reagent Reservoirs
GREENER_CHOICEBIOPHARMA

Product Code. 10261391 Shop All Corning Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
Packaging:
Bulk
Individually Wrapped
Unit Size:
Pack of 100
Pack of 200

Product Code. 10261391

Brand: Corning™ 4871

Only null left
Add to basket
Edge
Add to basket

Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.

Costar™ reagent reservoirs are ideal for repetitively filling multichannel pipettors.

  • Manufactured from modified polystyrene (white)
  • Individually wrapped, sterile
  • Disposable
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 mL
Dimensions (L x W x H) 145 x 70 x 28 mm
Material Polystyrene
Sterility Sterile
Height (Metric) 28 mm
Length (Metric) 145 mm
No. per Case 100/Cs.
No. per Pack 1/Pk.
Packaging Individually Wrapped
Width (Metric) 70 mm
Disposable Single-use
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.