Learn More
Corning™ Costar™ Sterile Disposable Reagent Reservoirs



Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.
Brand: Corning™ 4870
213.35 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.
Costar™ reagent reservoirs are ideal for repetitively filling multichannel pipettors.
- Manufactured from modified polystyrene (white)
- Individually wrapped, sterile
- Disposable

Specifications
50 mL | |
Polystyrene | |
Reagent Reservoir | |
145 mm | |
5/Pk. | |
70 mm |
145 x 70 x 28 mm | |
Sterile | |
28 mm | |
200/Cs. | |
Bulk | |
Single-use |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
4Hours
12Minutes
51Seconds
50
Your input is important to us. Please complete this form to provide feedback related to the content on this product.