Learn More
Corning™ Costar™ Thermowell™ Plate Lid
Fits styles C, M, and H.
Brand: Corning™ 6520
Code : 66
Additional Details : Weight : 0.40823kg
Description
- Polycarbonate lids are thermally stable up to 135°C and can be used on Corning™ Thermowell™ 96 well polycarbonate plates (fits Cat. No. 6511) with oil overlays
- Lids are intended to be covers only and do not prevent evaporation
Specifications
Polycarbonate | |
96 | |
1/Pk. | |
Thermowell Polycarbonate Plates |
Non-sterile | |
25/Cs. | |
Plate Lid |