missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Transwell™ Multiple Well Plate with Permeable Polyester Membrane Inserts
GREENER_CHOICEBIOPHARMA

Featuring a microscopically transparent polyester membrane that is TC-treated for optimal cell attachment and growth. Corning™ Transwell™ Multiwell Plate with Polyester Membrane Inserts provide better visibility under phase contrast microscopy and allow assessment of cell viability and monolayer formation.

Brand:  Corning™ 3452

 View more versions of this product

Product Code. 10699141

  • £215.00 / Pack of 24
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

 

  • Ideal for transport studies, chemotaxis, co-culture, and microbial pathogenesis studies
  • Thin membrane and straight pore structure provides consistent transport and cell migration
  • Self-centering insert prevents media from wicking between the insert and the outer well
  • Plate: Virgin Polystyrene
  • Membrane: PET

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 inserts in a 6-well plate
PET
2 x 106 pores/cm2
Well Plate
Packaged 6 inserts in a 6 well plate, 4 plates per case
TC-Treated
TC-Treated
6
4.67 cm2
2.6 mL
3 μm
24 mm
24 inserts, 4 plates/Case
Clear
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers