missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Transwell™-COL Collagen-Coated PTFE Membrane Insert
GREENER_CHOICE

Product Code. p-176731 Shop All Corning Products
Click to view available options
Pore Size:
0.4 μm
3 μm
For Use With (Equipment):
12 Well Plate
24 Well Plate
6 Well Plate
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 10042501

Brand: Corning™ undefined

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Transparent, collagen-treated microporous membranes promote cell attachment and spreading

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings Transparent, Collagen-coated PTFE membrane
  • Promotes cell attachment and spreading
  • Equimolar mixture of Collagen type I and III
  • Individually packaged
  • Multiple well plates included in each case
  • Sterile

Order Info

Individually packed and supplied in package with tissue culture plates with lids; sterile.

TRUSTED_SUSTAINABILITY

Specifications

Culture Area 0.33 cm2
Material PTFE
Volume (Metric) Well 0.6 mL
Pore Size 0.4 μm
Type Membrane Insert
For Use With (Equipment) 24 Well Plate
Diameter (Metric) Outer 6.5 mm
No. per Pack 1/Pk.
No. per Case 24/Cs.
Surface Coating Collagen
Sterility Sterile
No. of Wells 24
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.