missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZVG™ Cotton Cleaning Cloths
These cleaning cloths made of thin, strong cotton are absorbent, low-linting, and primarily suitable for polishing and cleaning tasks. Compliant with DIN 61650 standards.
Brand: ZVG™ 10097-01
This item is not returnable.
View return policy
Description
- 100% cotton
- Predominantly seamless
- Lint-free, absorbent material
Specifications
Cleaning Cloth | |
DIN 61650 | |
Single-use | |
Non-sterile |
Cotton | |
White | |
10 kg |