Learn More
ZVG™ Cotton Cleaning Cloths
These cleaning cloths made of thin, strong cotton are absorbent, low-linting, and primarily suitable for polishing and cleaning tasks. Compliant with DIN 61650 standards.
Brand:  ZVG™ 10098-01
 View more versions of this product
Product Code. 17873525
  • £41.05 / Each
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 100% cotton
  • Predominantly seamless
  • Lint-free, absorbent material
Specifications
Specifications
Cleaning Cloth
DIN 61650
Single-use
Non-sterile
Cotton
Multicolored
10 kg