Learn More
Sartorius™ Cotton Gloves for Weights
Prevent damage to the surface of calibration weights. Sartorius™ Cotton Gloves for Calibration Weights allow you to safely and properly handle finely machined calibration weights and maintain balance accuracy.
Brand: Sartorius™ YAW21
18.22 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 0.02700kg
Description
- Cotton construction
- Ensure accuracy of weight and mass standards
- Eliminates fingerprints and resists contamination by dirt, moisture, and corrosion
Specifications
Calibration Weights and Standards | |
White | |
Cotton | |
1 Pair/Pk. |