Learn More
Coulomat A, Honeywell™ Fluka™ HYDRANAL™
Brand: Honeywell Fluka 34807-500ML
89.05 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 67-56-1 Weight : 0.44050kg Transport : UN number : 1992 Chem class : 3 Pack group : II
Specifications
11°C (51.8°F) | |
HYDRANAL™-Coulomat A | |
59°C | |
UN3286 |
500 mL | |
1.070g/cm3 | |
Polycoated bottle |
Safety and Handling
- HYDRANAL-Coulomat A
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H331-Toxic if inhaled.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- H360D-May damage the unborn child.
- H351-Suspected of causing cancer.
- H370-Causes damage to organs.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : trichloromethane; chloroform, methanol,imidazole, sulphur dioxide, 1H-Imidazolemonohydriodide