missing translation for 'onlineSavingsMsg'
Learn More

Coumarin-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10525061
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-81-7
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.15
MDL Number MFCD00006852
InChI Key ACMLKANOGIVEPB-UHFFFAOYSA-N
Synonym coumarin-3-carboxylic acid, 2-oxo-2h-chromene-3-carboxylic acid, 3-carboxycoumarin, 2-oxobenzopyran-3-carboxylic acid, g 1 rodenticide, 2h-1-benzopyran-3-carboxylic acid, 2-oxo, 2-oxo-2h-1-benzopyran-3-carboxylic acid, unii-v85uov8788, g 1 the rodenticide van, g 1 van
PubChem CID 10752
IUPAC Name 2-oxochromene-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)C(=O)O

Specifications

Melting Point 190°C to 193°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 18, 429
Merck Index 15, 2551
Solubility Information Solubility in water: 13g/L (37°C). Other solubilities: soluble in alcohol and alkalies,insoluble in ether,benzene,petroleum ether
Formula Weight 190.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Coumarin-3-carboxylic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'