missing translation for 'onlineSavingsMsg'
Learn More
Coumarin 334, 96% Product Code.: 15476137

Coumarin 334, 96%

Product Code. 15476137
500 mg
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy

Product Code. 15476137

Brand: Thermo Scientific Alfa Aesar H33332.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55804-67-6 | C17H17NO3 | 283.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 55804-67-6
Melting Point 176°C to 181°C
Molecular Formula C17H17NO3
MDL Number MFCD00051336
Quantity 500 mg
UN Number UN2811
Synonym coumarin 334, 9-acetyl-2,3,5,6-tetrahydro-1h,4h-11-oxa-3a-aza-benzo de anthracen-10-one, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizin-11-one, 10-acetyl-2,3,6,7-tetrahydro, 10-acetyl-2,3,6,7-tetrahydro-1h,5h,11h-pyrano-2,3-f pyrido 3,2,1-ij quinolin-11-one, 10-acetyl-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinolin-11-one, coumarin-334, coumarin 521, coumarin 334, dye content 99 %, 3-acetyl-6,7,9,10-tetrahydro-8h,11h-quinolizino 9,9a, 1-gh coumarin
InChI Key JBPCDMSEJVCNGV-UHFFFAOYSA-N
SMILES CC(=O)C1=CC2=CC3=C4N(CCC3)CCCC4=C2OC1=O
IUPAC Name 5-acetyl-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Molecular Weight (g/mol) 283.33
PubChem CID 72655
Formula Weight 448.53
Percent Purity 96%
Chemical Name or Material Coumarin 334
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Coumarin 334, 96% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.