missing translation for 'onlineSavingsMsg'
Learn More

Coumarin, 99+%

Product Code. 10060790 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10060790

Brand: Thermo Scientific Acros 110530010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-64-5 | C9H6O2 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-64-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006850
InChI Key ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
Synonym 2H-1-Benzopyran-2-one
PubChem CID 323
ChEBI CHEBI:28794
IUPAC Name chromen-2-one
SMILES O=C1OC2=CC=CC=C2C=C1

Specifications

Melting Point 68°C to 71°C
Assay Percent Range 99+%
Beilstein 17, 328
Merck Index 15, 2550
Infrared Spectrum Authentic
Formula Weight 146.15
Percent Purity ≥99.0%
Packaging Plastic bottle
Color White
Quantity 1 kg
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Coumarin, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Coumarin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.