Learn More
Countess™ 3 21 CFR Part 11 Software Module
The Countess 3 21 CFR Part 11 Software Module is for use with the Countess 3 and Countess 3 FL automated cell counters and supports compliance with 21 CFR, Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using a SAE administration console.
Brand: Invitrogen A51025
Additional Details : Weight : 0.01000kg
Description
The Countess 3 21 CFR Part 11 Software Module is for use with the Countess 3 and Countess 3 FL automated cell counters and supports compliance with 21 CFR, Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using a SAE administration console.
21 CFR Part 11 compliance is composed of both procedural and technical requirements. Procedural requirements are the standard operating procedures instituted by the end user, and technical requirements are the functional characteristics of the compliance management software used. Together, when met, these requirements enable a customer to meet 21 CFR part 11 guidelines for traceability of electronic records.
To implement the Countess 3 21 CFR Part 11 Software Module for 21 CFR part 11 support on Countess 3 and Countess 3 FL automated cell counters, the following components are required to be installed, activated, and communicating:
- SAE Administrator Console—with the Countess 3 automated cell counter application profile, installed on a local server, used to configure the SAE settings for Countess 3 automated cell counters
- Countess 3 21 CFR part 11 license key—used to activate the SAE settings for the Countess 3 automated cell counter
- Countess 3 instrument, with SAE mode enabled—connected to the SAE Administrator Console
Features of the Countess 3 21 CFR Part 11 Software Module include:
Security
- Authentication of user login information
- Ability to set password guidelines and expiry dates
- Lockout access and re-authentication requirements after a period of inactivity
- System lockout upon unauthorized attempts to access software
- Ability of administrator to define user roles and permissions
- Verification of each user's permissions to perform predefined activities
- Identification of data tampering
Auditing
- Generation of audit trail log
- Tracking of changes and committed actions to user roles, account management, and system settings
- Ability of administrator to determine functions required to input reason for commit
- Audit trail record presented in readable and filterable formats
Electronic signature
- Administrator determination of e-signature requirements
- Ability to review records and sign electronically
Along with the 21 CFR part 11 software, our service and support organization offers IQ/OQ qualification services for your instrument and support training.
Specifications
Countess 3 & Countess 3 FL | |
21 CFR, part 11 activation key |