Learn More
Thermo Scientific™ Countess™ Light Cube Removal Tool
For use with Countess II FL Automated Cell Counter Only.
Brand: Thermo Scientific™ AMEP4747
Code : NEW
Additional Details : Weight : 1.00000kg
Description
For use with Countess II FL Automated Cell Counter Only. Used for installation or removal of EVOS Light Cubes from the Countess II FL instrument.
Specifications
Light Cube Remove Tool | |
Cell Counting | |
Room Temperature |
Countess™ | |
Countess II FL and 3 FL Automated Cell Counters |
For Research Use Only. Not for use in diagnostic procedures.