Learn More
Invitrogen™ Countess™ II FL Reusable Slide Holder
For use with Countess™ II FL Automated Cell Counter Only. Holds the Countess™ II FL Reusable Slide (A25750). Not compatible with Countess™ disposable slides.
Brand:  Invitrogen™ AMEP4746
Product Code. 15335656
  • £50.50 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Reusable Slide Holder
Cell Counting
1 unit
Countess
Countess™ Automated Cell Counter
Room Temperature