Learn More
Invitrogen™ Countess™ Reusable Slide
For use with the Countess™ II FL Automated Cell Counter Only. The Countess™ II FL Reusable Slide is a greener alternative to disposable slides.
Brand: Invitrogen™ A25750
274.88 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.50000kg
Specifications
Reusable Slide | |
Cell Counting | |
Room Temperature |
Countess™ | |
Countess II FL, Countess 3, and 3 FL Automated Cell Counters |