Learn More
Hirschmann™ Counting Chambers Double Net Ruling
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann™ Counting Chambers are available in a variety of depths to fit a range of systems.
Brand:  Hirschmann™ 8100111
 View more versions of this product
Product Code. 10521412
  • £186.00 / Each
Explore more special offers
Description
Description
  • Double net ruling
  • Including 2 cover glasses for hemacytometer
  • CE acc. IVD 98/79 EC
Certifications
CE acc. IVD 98/79 EC
Specifications
Specifications
Counting chambers, double net ruling
1 Each
2 cover glasses for haemacytometer
Counting chamber acc. to Nageotte
0.5 mm