Learn More
Hirschmann™ Cover Glasses for Microscopy
Preserve and protect specimens on slides. Hirschmann Instruments™ Cover Glasses for Microscopy are manufactured from quality glass and come in a range of dimensions.
£23.31 - £91.21
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10062481
|
Hirschmann™
8000102 |
18 x 18 mm |
£23.31
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10650193
|
Hirschmann™
8000105 |
24 x 24 mm |
£38.30
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13168159
|
Hirschmann™
8000110 |
22 x 40 mm |
£56.25
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13128159
|
Hirschmann™
8000104 |
22 x 22 mm |
£32.47
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13198159
|
Hirschmann™
8000115 |
24 x 32 mm |
£47.89
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13230480
|
Hirschmann™
8000119 |
24 x 50 mm |
£73.39
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13158159
|
Hirschmann™
8000109 |
22 x 32 mm |
£46.28
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10531942
|
Hirschmann™
8000103 |
20 x 20 mm |
£43.10
Pack of 1000 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
13178159
|
Hirschmann™
8000111 |
22 x 50 mm |
£71.95
Pack of 1000 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10165831
|
Hirschmann™
8000101 |
15 x 15 mm |
£23.75
Pack of 1000 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
13118169
|
Hirschmann™
8000117 |
24 x 46 mm |
£72.97
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13128169
|
Hirschmann™
8000118 |
24 x 48 mm |
£91.21
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13188159
|
Hirschmann™
8000112 |
22 x 60 mm |
£86.14
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- 0.13-0.17 mm thickness
- Acc. to DIN 58 884, CE acc. IVD 98/79 EC
Specifications
Coverslip | |
#1 (0.13 to 0.16 mm) |