Learn More
Epredia™ Cover Slips, Circles
Epredia™TM coverslips are ideally suited as cover material for specimens in microscopic examinations in medicine, biology and research. Manufactured from D 263TM M borosilicate glass from Schott AG. Available in various sizes thicknesses.
Brand:  Epredia™ CB00300RAC40MNZ0
 View more versions of this product
Product Code. 10343435
  • £258.00 / Pack of 1000
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Highly transparent and achromatic – practically free of any color distortion
    Good planarity
  • Refraction index adjusted for microscopes
  • High optical homogeneity – practically free of inclusions, streaks, bubbles etc.
  • Very good chemical stability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Circle
30 mm
#1.5 (0.16 to 0.19 mm)
Borosilicate Glass
30 mm
30 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ Cover Slips, Circles >