Learn More
Bollé Safety Coverall 2 Sterile Goggle
480.00 GBP valid until 2024-12-31
Use promo code "23287" to get your promotional price.
Use promo code "23287" to get your promotional price.
Click to view available options
Frame Color: Translucent
Description
- Can be sterilised using gamma, beta radiation and ETO gas
- Disposable
- Liquid overflow chute
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Frame Color | Translucent |
Includes | White Isoprene Strap |
Lens Material | Polycarbonate |
Frame Material | PVC |