Learn More
DuPont™ Tyvek 200 Easysafe Hooded Coverall
GREENER_CHOICE
Product Code. 15320541
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Offering adequate Type 5 and 6 barrier protection and remains both air and moisture vapour permeable. DuPont™ Tyvek™ 200 Easysafe Hooded Coveralls are made from a new and unique optimized polyethylene nonwoven that is softer with a more open structure than Tyvek™ 500 and 600.
  • Robust yet lightweight
  • Elasticated face, wrists, waist and ankles
  • Antistatic treatment on the outside
  • Stitched external seams in yellow for visual identification and differentiation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size Medium
Material Polyethylene
Color White
Ankle Style Elastic
Neck Style Hooded
Hood Type 2-panel
Closure Type Zipper
Seam Type Stitched
Protection Class Category III, Type 5 and 6
Certifications/Compliance (EU) 2016/425, EN 1073-2, EN 1149-1
Size (Metric) Chest 92 to 100 cm