Learn More
Surescreen Diagnostics COVID-19 Antigen Dual Test Cassette
COVID-19 Rapid Antigen Test for Saliva and Nasal fluid.
Brand: Surescreen Diagnostics COVID19AGVCD
This item is not returnable.
View return policy
Description
- For professional in vitro diagnostic use only – This product is CE-IVD marked for professional use and complies with the European In-Vitro Diagnostic Devices Directive (98/79/EC)
- A COVID-19 rapid test for the qualitative detection of COVID-19 in saliva and Nasopharyngeal swab
Specifications
Saliva/Nasopharyngeal Swab | |
Antigen Test | |
25 Tests/Kit |
Surescreen Antigen Dual Test CE
File size: 474 kB
EU Declaration of Conformity IVD Directive 98 97 EC
File size: 96.4 kB
Product Content Correction
Product Title
Surescreen Diagnostics COVID-19 Antigen Dual Test Cassette > 25 Tests/Kit
Spot an opportunity for improvement?Share a Content Correction