missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals COVID-19 Antigen Rapid Test (Rapid SARS-CoV-2 Antigen Test Card), 20 Tests/Kit

The COVID-19 Antigen Rapid Test (Rapid SARS-CoV-2 Antigen Test Card) is a rapid chromatographic immunoassay for the qualitative detection of SARS-CoV-2 antigens present in human nasal, nasopharyngeal, or oropharyngeal specimens.

Brand:  MP Biomedicals 07AG6020B

Product Code. 17140051

  • ÂŁ67.65 / 20 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The test is intended to aid in the rapid diagnosis of SARS-CoV-2 infections.

  • EASY TO USE: Easy sampling using a nasal, nasopharyngeal, or oropharyngeal swab specimen in only a few steps.
  • RAPID AND CONVENIENT: Results in 15 minutes. Requires no instruments.
  • EXCELLENT PERFORMANCE CHARACTERISTICS: Sensitivity 96.39%, Specificity 99.03%, Accuracy 98.59%

For professional in vitro diagnostic use only.

This product is CE-IVD marked for professional use and complies with the European In-Vitro Diagnostic Devices Directive (98/79/EC).

Specifications

Specifications

CE-IVD
Control included in each test strip
SARS-CoV-2 Nucleocapsid Protein Antigen
Immunochromatographic Assay
96.39%
Single-use
Swab, Buffer, Test Card and consumables for 20 tests
20 Tests/Kit
Kit
Human
SARS-CoV-2
2°C to 30°C
Yes
No
Nasal, Nasopharyngeal, or Oropharyngeal swab specimens
Rapid Test
Lateral Flow Rapid Test
Plastic Card
Human
0.3 kg
99.03%
15 min.
Product Suggestions

Product Suggestions

Videos

Videos

  • MP Biomedicals Rapid SARS-CoV-2 Antigen Test Card
    MP Biomedicals Rapid SARS-CoV-2 Antigen Test Card
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      MP Biomedicals Rapid SARS-CoV-2 Antigen Test Card

    SDS
    Documents

    Documents

    Certificates
    View accessory products
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    MP Biomedicals COVID-19 Antigen Rapid Test (Rapid SARS-CoV-2 Antigen Test Card), 20 Tests/Kit > 20 Tests/Kit

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.