missing translation for 'onlineSavingsMsg'
Learn More

CP 94253 hydrochloride, Tocris Bioscience™

Product Code. 30001233
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective 5-HT1B agonist

Potent, selective 5-HT1B agonist (Ki values are 89, 2, 860, 49 and 1,600 nM for 5-HT1A, 5-HT1B, 5-HT1C, 5-HT1D and 5-HT2 receptors respectively). Centrally active upon systemic administration in vivo.

Chemical Identifiers

CAS 845861-39-4
Molecular Formula C15H20ClN3O
Molecular Weight (g/mol) 293.795
InChI Key PIIOXKQIZCVXMD-UHFFFAOYSA-N
Synonym cp 94253 hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridine hydrochloride, 5-propoxy-3-1,2,3,6-tetrahydropyridin-4-yl-1h-pyrrolo 3,2-b pyridine hydrochloride, zlchem 1209, 5-propoxy-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-pyrrolo 3,2-b pyridinehydrochloride, cp 94253 hydrochloride/, 4-5-propoxy-1h-pyrrolo 3,2-b pyridin-3-yl-1,2,3,6-tetrahydropyridine hydrochloride
PubChem CID 11652258
IUPAC Name 5-propoxy-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-pyrrolo[3,2-b]pyridine;hydrochloride
SMILES CCCOC1=NC2=C(C=C1)NC=C2C3=CCNCC3.Cl

Specifications

Quantity 10 mg
Formula Weight 293.8
Percent Purity >98%
Chemical Name or Material CP 94253 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'